missing translation for 'onlineSavingsMsg'
Learn More

Sodium Perborate Tetrahydrate, ≥10% Active Oxygen Basis, Honeywell™

Product Code. p-7120818 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Poly bottle
Unit Size:
1kg
250g
Product Code. Quantity Packaging unitSize
15666850 250 g Poly bottle 250g
15656850 1 kg Poly bottle 1kg
2 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 15666850

Brand: Honeywell Research Chemicals 71840250G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Purum p.a.,≥97.0% (RT)

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10486-00-7
MDL Number MFCD00149231

Specifications

Boiling Point Decomposes on heating
Melting Point 60°C
Color White
Physical Form Crystalline Powder
Quantity 250 g
Linear Formula NaBO3 · 4H2O
UN Number UN1479
Solubility Information Soluble
Formula Weight 153.86g/mol
Percent Purity ≥10% (active oxygen basis)
Odor Odorless
Packaging Poly bottle
Chemical Name or Material Sodium perborate tetrahydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.