Learn More
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Specifications
Specifications
| Melting Point | 60.0°C |
| Color | White |
| pH | 10.1 to 10.6 (1% soln.) |
| Physical Form | Crystalline Powder |
| Quantity | 1 kg |
| Assay Percent Range | 97% |
| Linear Formula | NaBO3·4H2O |
| Fieser | 01,1102; 02,387; 04,59; 14,290; 16,310 |
| Merck Index | 14,8652 |
| Solubility Information | Solubility in water: 21.5g/L (18°C) |
| Show More |
Safety and Handling
- Sodium perborate tetrahydrate
- Danger
- Serious eye damage/eye irritation Category 1
- Reproductive toxicity Category 1B
- Specific target organ toxicity Category 3
- Acute toxicity Category 4
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
- H360Df-May damage the unborn child. Suspected of damaging fertility.
- H332-Harmful if inhaled.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Sodium perborate tetrahydrate
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.