missing translation for 'onlineSavingsMsg'
Learn More

Sodium perborate tetrahydrate, 97%

Product Code. 11443970
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
Product Code. Quantity unitSize
11443970 100 g 100g
11409502 500 g 500g
11419502 2500 g 2500g
3 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 11443970

Brand: Thermo Scientific Alfa Aesar A10189.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

CAS: 10486-00-7 | NaBO3·4H2O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Sodium perborate tetrahydrate is considered as a source of active hydrogen used in detergents, cleaning products, laundry detergents and bleaches. It finds application as a tooth bleaching agent utilized for tooth. It has antiseptic properties and acts as disinfectant as well as preservative in eye drops. Further, it is mixed with suitable activator, tetraacetylethylenediamine in order to release oxygen at lower temperatures.

Solubility
Soluble in water.

Notes
Moisture sensitive.Incompatible with metals, metallic salts, acids, bases and reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10486-00-7
Molecular Formula NaBO3·4H2O
MDL Number MFCD00149231

Specifications

Melting Point 60°C (decomposition)
Density 1.73
pH ∼10
Odor Odorless
Quantity 100 g
UN Number UN1479
Sensitivity Hygroscopic
Merck Index 14,8652
Solubility Information Soluble in water.
Formula Weight 153.86 (81.84 Anhydrous)
Percent Purity 97%
Chemical Name or Material Sodium perborate tetrahydrate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.