missing translation for 'onlineSavingsMsg'
Learn More

Sodium Perborate Tetrahydrate, Extra Pure, SLR, Fisher Chemical

Product Code. 10755151 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Plastic powder jar
Unit Size:
500g
Product Code. Quantity Packaging unitSize
10755151 500 g Plastic powder jar 500g
1 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 10755151

Brand: Fisher Chemical S/5880/53

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Specifications

Melting Point 60°C
CAS 10486-00-7
Color White
pH 10.4
Physical Form Powder/Solid
Quantity 500 g
Molecular Formula BNaO3·4H2O
MDL Number 149231
Formula Weight 153.86g/mol
Packaging Plastic powder jar
Vapor Pressure 1.3mbar 30
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Sodium perborate tetrahydrate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Acute toxicity Category 4
  • Specific target organ toxicity Category 3
  • Reproductive toxicity Category 1B
Hazard Statement
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
Precautionary Statement
  • P202-Do not handle until all safety precautions have been read and understood.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Supplemental information
  • MIXTURE LIST-Contains : Sodium perborate tetrahydrate, Sodium perborate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.