missing translation for 'onlineSavingsMsg'
Learn More

Sodium perchlorate, 99+%, for analysis

Product Code. 11994211 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
250 g
1 kg
5 kg
Packaging:
Glass Bottle
Plastic bottle
Conditionnement:
1kg
250g
5g
5kg
Code produit. Quantity Packaging unitSize
11994211 5 kg Plastic bottle 5kg
10032570 5 g Glass Bottle 5g
10752241 250 g Plastic bottle 250g
10012560 1 kg Plastic bottle 1kg
4 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Sea freight only: For EU stock allow up to 15 days delivery.
Règlementaire: Ce produit contient un précurseur d'explosif selon le règlement (CE) n ° 1148/2019, tout client doit remplir une déclaration avant la livraison.

Code produit. 11994211

Marque: Thermo Scientific Acros 197120051

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Sea freight only: For EU stock allow up to 15 days delivery.
Règlementaire: Ce produit contient un précurseur d'explosif selon le règlement (CE) n ° 1148/2019, tout client doit remplir une déclaration avant la livraison.

CAS: 7601-89-0 | ClNaO4 | 122.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 7601-89-0
InChI Key BAZAXWOYCMUHIX-UHFFFAOYSA-M
Synonym sodium perchlorate, perchloric acid, sodium salt, natriumperchlorat, perchlorate de sodium, natriumperchloraat dutch, natriumperchlorat german, unii-97f4mty3va, cpl 46, perchlorate de sodium french, sodio perclorato di italian
PubChem CID 522606
IUPAC Name sodium;perchlorate
SMILES [O-]Cl(=O)(=O)=O.[Na+]

Spécification

Melting Point 482.0°C
Color White
pH 6.0 to 8.0 (5% aq. soln.)
Physical Form Crystalline Powder and/or Crystals
Quantity 5 kg
Assay Percent Range 99% min. (on dry substance)
Molecular Formula ClNaO4
Linear Formula NaClO4
Merck Index 15,8784
Solubility Information Solubility in water: 209 g/100mL (15°C). Other solubilities: 170 g/100mL water (0°C), 300 g/100mL water (80°C), soluble in alcohol
Molecular Weight (g/mol) 122.44
Formula Weight 122.44
Percent Purity ≥99%
Grade Analytical
Loss on Drying 2.0% max. (130°C, 2 hrs)
Packaging Plastic bottle
Chemical Name or Material Sodium perchlorate
Afficher plus Afficher moins
compliance-icons compliance-icons compliance-icons
Identifiant produit
  • Sodium perchlorate
Mention d'avertissement
  • Danger
Catégorie de danger
  • Oxidising solid Category 1
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
Mention de danger
  • H271-May cause fire or explosion; strong oxidiser.
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
  • H373-May cause damage to organs.
Conseils de prudence
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P371+P380+P375-In case of major fire and large quantities: Evacuate area Fight fire remotely due to the risk of explosion.

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.