missing translation for 'onlineSavingsMsg'
Learn More

Sodium perchlorate, anhydrous, ACS, 98.0-102.0%

Product Code. 11381718
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
Product Code. Quantity unitSize
11381718 100 g 100g
11371718 500 g 500g
2 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Product Code. 11381718

Brand: Thermo Scientific Alfa Aesar 011623.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

CAS: 7601-89-0 | ClNaO4 | 122.436 g/mol

Sodium perchlorate is used in chemical analysis and in explosive materials, fireworks, and rocket motors. It is the precursor for the preparation of other perchlorate salts. It is also employed as a chaotrophic agent in DNA extraction and hybridization reactions in molecular biology. It prevents the binding of phenolic compounds to RNA during extraction. In addition, it is used in batteries, plastics and in rubber products. It is also used as a nonreactive electrolyte. Further, it is an eluent component used for analysis of biological samples by liquid chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7601-89-0
Molecular Formula ClNaO4
Molecular Weight (g/mol) 122.436
MDL Number MFCD00003480
InChI Key BAZAXWOYCMUHIX-UHFFFAOYSA-M
Synonym sodium perchlorate, perchloric acid, sodium salt, natriumperchlorat, perchlorate de sodium, natriumperchloraat dutch, natriumperchlorat german, unii-97f4mty3va, cpl 46, perchlorate de sodium french, sodio perclorato di italian
PubChem CID 522606
IUPAC Name sodium;perchlorate
SMILES [O-]Cl(=O)(=O)=O.[Na+]

Specifications

Melting Point 482°C (decomposition)
Physical Form Crystalline
Quantity 100 g
Assay Percent Range 98.0 to 102.0%
Solubility Information Soluble in water and alcohol.
Formula Weight 122.46
Grade ACS Reagent
Sensitivity Hygroscopic
Density 2.50 g/mL
Chemical Name or Material Sodium perchlorate, anhydrous, For analysis ACS
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Sodium perchlorate, anhydrous
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Oxidising solid Category 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H271-May cause fire or explosion; strong oxidiser.
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
  • H373-May cause damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P371+P380+P375-In case of major fire and large quantities: Evacuate area Fight fire remotely due to the risk of explosion.
Supplemental information
  • MIXTURE LIST-Contains : Sodium perchlorate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.