missing translation for 'onlineSavingsMsg'
Learn More

Sodium Perchlorate, Certified AR for Analysis, Fisher Chemical

Product Code. 10775531 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Plastic powder jar
Unit Size:
500g
Product Code. Quantity Packaging unitSize
10775531 500 g Plastic powder jar 500g
1 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Product Code. 10775531

Brand: Fisher Chemical S/5965/53

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Specifications

Melting Point 130°C
CAS 7-3-7791
Color White
pH 6.0 to 8.0
Physical Form Solid
Quantity 500 g
Molecular Formula ClH2NaO5
MDL Number 149164
Synonym sodium perchlorate monohydrate, sodium perchlorate hydrate, unii-g4h485lhcw, g4h485lhcw, ccris 8893, perchloric acid, sodium salt, monohydrate, sodium hydrate perchlorate, clo4.na.h2o, acmc-1bii1, sodium perchlorate-monohydrate
InChI Key IXGNPUSUVRTQGW-UHFFFAOYSA-M
SMILES O.[O-]Cl(=O)(=O)=O.[Na+]
IUPAC Name sodium;perchlorate;hydrate
Molecular Weight (g/mol) 140.45
PubChem CID 516899
Formula Weight 140.46g/mol
Packaging Plastic powder jar
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Sodium perchlorate monohydrate
Signal Word
  • Danger
Hazard Category
  • Oxidising solid Category 1
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H271-May cause fire or explosion; strong oxidiser.
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
  • H373-May cause damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P371+P380+P375-In case of major fire and large quantities: Evacuate area Fight fire remotely due to the risk of explosion.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.