missing translation for 'onlineSavingsMsg'
Learn More

Sodium Perchlorate, for HPLC, Thermo Scientific
GREENER_CHOICE

Product Code. 30280942 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30280942 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30280942 Supplier Thermo Scientific Supplier No. T059660250
  • ONLINE EXCLUSIVE PRICE
    125.00 €
    113.00 € / 250g
    Save 12.00 €
    9% Off
View all promotions
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

Specifications

CAS 7-3-7791
Physical Form Powder, Crystals and/or Chunks
Quantity 250 g
Molecular Weight (g/mol) 140.45 g/mol
Percent Purity 99%
Grade HPLC
Packaging Plastic Powder Jar
Chemical Name or Material Sodium Perchlorate
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Sodium perchlorate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Oxidising solid Category 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H271-May cause fire or explosion; strong oxidiser.
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
  • H373-May cause damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P314-Get medical advice/attention if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Sodium perchlorate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.