missing translation for 'onlineSavingsMsg'
Learn More

Sodium Perfluoro-1-nonanesulfonate (50 ug/mL in Methanol), TRC

Product Code. 30478106
Change view
Click to view available options
Quantity:
1x10 mL
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30478106 1x10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30478106 Supplier Toronto Research Chemicals Supplier No. TRCKIT81601X10ML
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Perfluorononanesulfonic acid sodium salt
CAS 98789-57-2
Molecular Formula C9 F19 O3 S . Na
Quantity 1x10 mL
Synonym 1-Nonanesulfonic acid, 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-nonadecafluoro-, sodium salt (1:1), 1-Nonanesulfonic acid, 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-nonadecafluoro-, sodium salt (9CI), Sodium 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-nonadecafluoro-1-nonanesulfonate (1:1), Sodium perfluorononanesulfonate
InChI Formula InChI=1S/C9HF19O3S.Na/c10-1(11,2(12,13)4(16,17)6(20,21)8(24,25)26)3(14,15)5(18,19)7(22,23)9(27,28)32(29,30)31;/h(H,29,30,31);/q;+1/p-1
SMILES [Na+].[O-]S(=O)(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
IUPAC Name sodium;1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-nonadecafluorononane-1-sulfonate
Molecular Weight (g/mol) 572.12
Formula Weight 571.9162
Concentration or Composition (by Analyte or Components) 50 μg/mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.