missing translation for 'onlineSavingsMsg'
Learn More

Sodium perfluorooctanoate, 97%, Thermo Scientific™

Product Code. 11323886
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11323886 5 g 5g
11333886 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11323886 Supplier Thermo Scientific Alfa Aesar Supplier No. L16988.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Sodium perfluorooctanoate on gel-to-liquid-crystalline phase transition of dipalmitoylphosphatidylcholine vesicle membrane.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 335-95-5
Molecular Formula C8F15NaO2
Molecular Weight (g/mol) 436.052
MDL Number MFCD00040394
InChI Key LWHQXUODFPPQTL-UHFFFAOYSA-M
Synonym sodium perfluorooctanoate, sodium pentadecafluorooctanoate, sodium perfluorocaprylate, octanoic acid, pentadecafluoro-, sodium salt, perfluorooctanoic acid sodium salt, unii-63a7smf12m, pentadecafluorooctanoic acid, sodium salt, sodium pentadecafluoro octanoate, octanoic acid, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluoro-, sodium salt 1:1, potassium pentadecafluorooctanoate
PubChem CID 23667657
IUPAC Name sodium;2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctanoate
SMILES C(=O)(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)[O-].[Na+]

Specifications

Melting Point ∽275°C (decomposition)
Linear Formula CF3(CF2)6CO2Na
Quantity 5 g
Solubility Information Soluble in water.
Formula Weight 436.05
Percent Purity 97%
Chemical Name or Material Sodium perfluorooctanoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.