missing translation for 'onlineSavingsMsg'
Learn More

Sodium Periodate, Solstice

Product Code. 15693270 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Unit Size:
100g
2.5kg
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15693270 100 g 100g
15623280 5 g 5g
15613280 500 g 500g
15603280 2.5 kg 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15693270 Supplier Honeywell-Fluka Supplier No. 311448100G
  • ONLINE EXCLUSIVE PRICE
    170.00 €
    153.65 € / 100g
    Save 16.35 €
    9% Off
 Restricted Item
Request Bulk or Custom Format
Currently Unavailable: We have no information when this product will be available.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

ACS Reagent,≥99.8%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-28-5
Molecular Formula INaO4
Molecular Weight (g/mol) 213.89
MDL Number MFCD00003534
InChI Key JQWHASGSAFIOCM-UHFFFAOYSA-M
Synonym sodium periodate, sodium metaperiodate, sodium m-periodate, sodiumperiodate, sodium meta periodate, periodic acid, sodium salt, sodium meta-periodate, periodic acid hio4 , sodium salt, periodate sodium, sodium penodate
PubChem CID 23667635
ChEBI CHEBI:75226
IUPAC Name sodium;periodate
SMILES [Na+].[O-][I](=O)(=O)=O

Specifications

Quantity 100 g
Linear Formula NaIO4
UN Number 3085
Formula Weight 213.89g/mol
Percent Purity ≥99.8%
Grade ACS Reagent
Chemical Name or Material Sodium Periodate
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Sodium periodate
Signal Word
  • Danger
Hazard Category
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Oxidising solid Category 1
  • Skin corrosion/irritation Category 1 C
  • Specific target organ toxicity Category 1
Hazard Statement
  • H271-May cause fire or explosion; strong oxidiser.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Sodium periodate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.