missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Peroxide, Extra Pure, SLR, Granulated, Fisher Chemical
Tienda Fisher Chemical Productos
Click to view available options
Quantity:
500 g
Packaging:
Plastic powder jar
Tamaño de la unidad:
500g
Especificaciones
Especificaciones
| Melting Point | 460°C |
| CAS | 1313-60-6 |
| Color | Yellow |
| Physical Form | Solid |
| Quantity | 500 g |
| Molecular Formula | Na2O2 |
| MDL Number | 3497 |
| Synonym | sodium peroxide, disodium peroxide, solozone, natriumsuperoxid, sodium dioxide, disodium dioxide, natrium peroxydatum, sodium oxide, per, flocool 180, sodium oxide |
| InChI Key | PFUVRDFDKPNGAV-UHFFFAOYSA-N |
| SMILES | [O-][O-].[Na+].[Na+] |
| Mostrar más |
Seguridad y manipulación
Nombre del producto
- Sodium peroxide
- Danger
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Oxidising solid Category 1
- Skin corrosion/irritation Category 1 A
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H290-May be corrosive to metals.
- H271-May cause fire or explosion; strong oxidiser.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido