missing translation for 'onlineSavingsMsg'
Learn More

Sodium peroxodisulfate, (sodiumpersulfate), Solstice

Product Code. 16605743
Change view
Click to view available options
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
16605743 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16605743 Supplier Fluka Supplier No. 134571KG
 Restricted Item
Request Bulk or Custom Format
Currently Unavailable: We have no information when this product will be available.
View alternative products
This item is not returnable. View return policy

Specifications

CAS 7775-27-1
Color Colorless
CAS Max % 1
pH 2.5 to 4
Quantity 1 kg
Molecular Formula Na2O8S2
InChI Key CHQMHPLRPQMAMX-UHFFFAOYSA-L
SMILES [Na+].[Na+].[O-]S(=O)(=O)OOS([O-])(=O)=O
IUPAC Name disodium [(sulfonatoperoxy)sulfonyl]oxidanide
Molecular Weight (g/mol) 238.09
Grade Reagent
Chemical Name or Material Sodium peroxodisulfate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.