missing translation for 'onlineSavingsMsg'
Learn More

Sodium peroxydisulfate, 98%

Product Code. 11438660
Change view
Click to view available options
Quantity:
1 kg
5 kg
Unit Size:
1kg
5kg
Product Code. Quantity unitSize
11438660 5 kg 5kg
11448660 1 kg 1kg
2 options
This item is not returnable. View return policy

Product Code. 11438660

Brand: Thermo Scientific Alfa Aesar 054100.A7

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7775-27-1 | Na2O8S2 | 238.092 g/mol

Sodium peroxydisulfate is used as a radical initiator for emulsion polymerization reactions like acrylonitrile butadiene styrene, detergent component, soil conditioner and soil remediation. It is also used for curing of formaldehyde adhesives. It acts as a bleaching agent and in the production of dyestuffs. It finds application in zinc and printed circuit boards and considered to be a replacement for ammonium persulfate in etching mixtures. Further, it is used in the preparation of diapocynin from apocynin. It plays a role in the conversion of phenols to para-diphenols in alkaline solution and of arylamines to aminophenols. It is actively involved in the Elbs persulfate oxidation and the Boyland-Sims oxidation reactions as an oxidizing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7775-27-1
Molecular Formula Na2O8S2
Molecular Weight (g/mol) 238.092
MDL Number MFCD00003501
InChI Key CHQMHPLRPQMAMX-UHFFFAOYSA-L
Synonym sodium persulfate, sodium peroxydisulfate, sodium peroxodisulfate, peroxydisulfuric acid, disodium salt, disodium peroxodisulphate, sodium persulphate, unii-j49fyf16je, persulfate de sodium french, sodium peroxydisulphate, disodium sulfonatooxy sulfate
PubChem CID 62655
IUPAC Name disodium;sulfonatooxy sulfate
SMILES [O-]S(=O)(=O)OOS(=O)(=O)[O-].[Na+].[Na+]

Specifications

Physical Form Crystalline
Quantity 5 kg
Assay Percent Range 98%
Solubility Information Soluble in water.
Formula Weight 238.1
Density 2.40 g/mL
Chemical Name or Material Sodium peroxydisulfate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.