missing translation for 'onlineSavingsMsg'
Learn More

Sodium Persulfate, >98% (White Crystalline Powder), Fisher BioReagents

Product Code. 11727499
Change view
Click to view available options
Quantity:
1 kg
Packaging:
Poly Bottle
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11727499 1 kg Poly Bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11727499 Supplier Fisher BioReagents Supplier No. BP26371

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Sodium Persulfate is used as a promoter for polymerization reactions.
TRUSTED_SUSTAINABILITY

Specifications

Melting Point 100°C
CAS 7775-27-1
Color White
pH 5 to 7
Physical Form Powder/Solid
Quantity 1 kg
Assay Percent Range ≥98.0 %
Molecular Formula Na2O8S2
Merck Index 15, 8787
Synonym sodium persulfate, sodium peroxydisulfate, sodium peroxodisulfate, peroxydisulfuric acid, disodium salt, disodium peroxodisulphate, sodium persulphate, unii-j49fyf16je, persulfate de sodium french, sodium peroxydisulphate, disodium sulfonatooxy sulfate
InChI Key CHQMHPLRPQMAMX-UHFFFAOYSA-L
SMILES [O-]S(=O)(=O)OOS(=O)(=O)[O-].[Na+].[Na+]
IUPAC Name disodium;sulfonatooxy sulfate
Molecular Weight (g/mol) 238.092
PubChem CID 62655
Formula Weight 238.09
Percent Purity ≥98.0%
Identification Pass Test
Packaging Poly Bottle
Density 2.6g/cm³
Chemical Name or Material Sodium Persulfate, >98%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Sodium Persulfate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Oxidising solid Category 3
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H272-May intensify fire; oxidiser.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.