missing translation for 'onlineSavingsMsg'
Learn More

Sodium persulfate, 99.5%, MP Biomedicals™

Product Code. 11485442 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11485442 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11485442 Supplier MP Biomedicals Supplier No. 0219549991
 Restricted Item
Currently Unavailable: We have no information when this product will be available.
View alternative products
This item is not returnable. View return policy

Specifications

CAS 7775-27-1
Color White
Physical Form Crystalline Powder
Quantity 1 kg
Assay Percent Range 0.995
Molecular Formula Na2O8S2
Synonym sodium persulfate, sodium peroxydisulfate, sodium peroxodisulfate, peroxydisulfuric acid, disodium salt, disodium peroxodisulphate, sodium persulphate, unii-j49fyf16je, persulfate de sodium french, sodium peroxydisulphate, disodium sulfonatooxy sulfate
InChI Key CHQMHPLRPQMAMX-UHFFFAOYSA-L
SMILES [O-]S(=O)(=O)OOS(=O)(=O)[O-].[Na+].[Na+]
IUPAC Name disodium;sulfonatooxy sulfate
Molecular Weight (g/mol) 238.092
PubChem CID 62655
Formula Weight 238.13
Density 1250 - 1300kg/m3 (bulk density)
Chemical Name or Material Sodium persulfate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.