missing translation for 'onlineSavingsMsg'
Learn More

Sodium Persulfate, Honeywell

Product Code. 15632030 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
5 kg
Unit Size:
1kg
2.5kg
500g
5kg
Product Code. Quantity unitSize
15632030 500 g 500g
15602030 1 kg 1kg
15612030 2.5 kg 2.5kg
15642030 5 kg 5kg
4 options
This item is not returnable. View return policy

Product Code. 15632030

Brand: Honeywell Research Chemicals 216232500G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Reagent Grade,≥98%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7775-27-1
Molecular Formula Na2O8S2
Molecular Weight (g/mol) 238.092
MDL Number MFCD00003501
InChI Key CHQMHPLRPQMAMX-UHFFFAOYSA-L
Synonym sodium persulfate, sodium peroxydisulfate, sodium peroxodisulfate, peroxydisulfuric acid, disodium salt, disodium peroxodisulphate, sodium persulphate, unii-j49fyf16je, persulfate de sodium french, sodium peroxydisulphate, disodium sulfonatooxy sulfate
PubChem CID 62655
IUPAC Name disodium;sulfonatooxy sulfate
SMILES [O-]S(=O)(=O)OOS(=O)(=O)[O-].[Na+].[Na+]

Specifications

Quantity 500 g
Linear Formula Na2S2O8
UN Number 1505
Formula Weight 238.1g/mol
Percent Purity ≥98%
Grade Reagent
Chemical Name or Material Sodium Persulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.