missing translation for 'onlineSavingsMsg'
Learn More

Sodium Persulfate, MP Biomedicals

Product Code. 11475442 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11475442 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11475442 Supplier MP Biomedicals Supplier No. 0219549990
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

CAS 7775-27-1
Quantity 500 g
Molecular Formula Na2O8S2
Synonym sodium persulfate, sodium peroxydisulfate, sodium peroxodisulfate, peroxydisulfuric acid, disodium salt, disodium peroxodisulphate, sodium persulphate, unii-j49fyf16je, persulfate de sodium french, sodium peroxydisulphate, disodium sulfonatooxy sulfate
InChI Key CHQMHPLRPQMAMX-UHFFFAOYSA-L
SMILES [O-]S(=O)(=O)OOS(=O)(=O)[O-].[Na+].[Na+]
IUPAC Name disodium;sulfonatooxy sulfate
Molecular Weight (g/mol) 238.092
PubChem CID 62655
Formula Weight 238.1
Chemical Name or Material Sodium Persulfate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.