missing translation for 'onlineSavingsMsg'
Learn More

Sodium Phosphate, Dibasic, 7-Hydrate, Crystal, U.S.P., J.T. Baker™

Product Code. 15558174 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Wide Mouth Amber Glass Bottle
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15558174 500 g Wide Mouth Amber Glass Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15558174 Supplier Avantor J.T.Baker Supplier No. 381701
  • ONLINE EXCLUSIVE PRICE
    318.84 €
    286.65 € / 500g
    Save 32.19 €
    10% Off
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7782-85-6 | Formula: H15Na2O11P | Weight: 268.06 g/mol

Chemical Identifiers

CAS 7782-85-6
Molecular Formula H15Na2O11P
Molecular Weight (g/mol) 268.06
MDL Number MFCD00149180
InChI Key LVXHNCUCBXIIPE-UHFFFAOYSA-L
Synonym sodium phosphate dibasic heptahydrate, unii-70wt22sf4b, sodium phosphate, dibasic, heptahydrate, sodium hydrogen phosphate heptahydrate, disodium hydrogen phosphate heptahydrate, axea paragraph signthornaae, 2na.hpo4.7h2o, sodium hydrogenphosphate heptahydrate, sodium phosphate dibasic, acs, disodium hydrogenphosphate heptahydrate
PubChem CID 6096963
IUPAC Name disodium phosphoric acid heptahydrate
SMILES O.[Na+].[Na+].OP([O-])([O-])=O

Specifications

Physical Form Crystals
Quantity 500 g
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 1.70kg
Formula Weight 268.07
Percent Purity 98.0 to 101.0%
Grade USP
Chemical Name or Material Sodium Phosphate, Dibasic, 7-Hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.