missing translation for 'onlineSavingsMsg'
Learn More

Sodium phosphate, dibasic, 98+%, extra pure, anhydrous

Product Code. 12685147 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
5 kg
25 kg
Unit Size:
1kg
25kg
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12685147 25 kg 25kg
12665147 1 kg 1kg
12675147 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 12685147 Supplier Thermo Scientific Acros Supplier No. 448140250
  • ONLINE EXCLUSIVE PRICE
    755.00 €
    731.00 € / 25kg
    Save 24.00 €
    3% Off
View all promotions
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 7558-79-4 | HNa2O4P | 141.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7558-79-4
Molecular Formula HNa2O4P
Molecular Weight (g/mol) 141.96
MDL Number MFCD00003496
InChI Key BNIILDVGGAEEIG-UHFFFAOYSA-L
Synonym disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt
PubChem CID 24203
ChEBI CHEBI:34683
SMILES [Na+].[Na+].OP([O-])([O-])=O

Specifications

Quantity 25 kg
Assay Percent Range 98%
Linear Formula Na2HPO4
Merck Index 15,8789
Solubility Information Solubility in water: 75g/L (20°C). Other solubilities: practically insoluble in alcohol
IUPAC Name disodium hydrogen phosphate
Formula Weight 141.96
Percent Purity ≥98%
Grade Extra Pure
Chemical Name or Material Sodium phosphate, dibasic

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.