missing translation for 'onlineSavingsMsg'
Learn More

Sodium phosphate dibasic, ACS Reagent, Solstice

Product Code. 15688160 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
2.5 kg
5 kg
6 x 500 g
12 kg
4 x 2.5 kg
Unit Size:
100g
12kg
1kg
2.5kg
500g
5kg
Pack of 4
Pack of 6
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15688160 500 g 500g
15638160 100 g 100g
15658160 1 kg 1kg
15668160 2.5 kg 2.5kg
15608170 6 x 500 g Pack of 6
15698160 5 kg 5kg
15678160 4 x 2.5 kg Pack of 4
15648160 12 kg 12kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Product Code. 15688160 Supplier Honeywell-Fluka Supplier No. S9763500G
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

ACS Reagent,≥99.0%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7558-79-4
Molecular Formula HNa2O4P
Molecular Weight (g/mol) 141.96
MDL Number MFCD00003496
InChI Key BNIILDVGGAEEIG-UHFFFAOYSA-L
Synonym disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt
PubChem CID 24203
ChEBI CHEBI:34683
SMILES [Na+].[Na+].OP([O-])([O-])=O

Specifications

Physical Form Powder
Quantity 500 g
Linear Formula Na2HPO4
UN Number NONH for all modes of transport
IUPAC Name disodium;hydrogen phosphate
Formula Weight 141.96g/mol
Percent Purity ≥99%
Grade Not Available
Chemical Name or Material Sodium phosphate dibasic
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.