missing translation for 'onlineSavingsMsg'
Learn More

Sodium phosphate dibasic, ACS Reagent, Honeywell Fluka™

Product Code. p-7072323 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
100 g
500 g
1 kg
2.5 kg
5 kg
6 x 500 g
12 kg
4 x 2.5 kg
Unit Size:
100g
12kg
1kg
2.5kg
500g
5kg
Pack of 4
Pack of 6
This item is not returnable. View return policy

Product Code. 15638160

Brand: Honeywell-Fluka S9763100G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ACS Reagent,≥99.0%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7558-79-4
Molecular Formula HNa2O4P
Molecular Weight (g/mol) 141.96
MDL Number MFCD00003496
InChI Key BNIILDVGGAEEIG-UHFFFAOYSA-L
Synonym disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt
PubChem CID 24203
ChEBI CHEBI:34683
SMILES [Na+].[Na+].OP([O-])([O-])=O

Specifications

Quantity 100 g
Linear Formula Na2HPO4
UN Number NONH for all modes of transport
IUPAC Name disodium;hydrogen phosphate
Formula Weight 141.96g/mol
Percent Purity ≥99%
Grade ACS Reagent
Chemical Name or Material Sodium phosphate dibasic
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.