missing translation for 'onlineSavingsMsg'
Learn More

Sodium Phosphate Dibasic Anhydrous (White Granules or Powder), Fisher BioReagents™

Product Code. 10396313
Change view
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Glass Bottle
Poly Bottle
Unit Size:
1kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10396313 1 kg Poly Bottle 1kg
10182863 500 g Glass Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10396313 Supplier Fisher BioReagents Supplier No. BP3321
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Sodium Phosphate Dibasic Anhydrous (White Granules or Powder)

This buffer component is commonly used in tissue culture and molecular biology applications.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7558-79-4
Molecular Formula HNa2O4P
Molecular Weight (g/mol) 141.96
MDL Number MFCD00003496
InChI Key BNIILDVGGAEEIG-UHFFFAOYSA-L
Synonym disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt
PubChem CID 24203
ChEBI CHEBI:34683
SMILES [Na+].[Na+].OP([O-])([O-])=O

Specifications

Color White
pH 8.7 to 9.3
Physical Form Powder/Solid
Quantity 1 kg
Assay Percent Range ≥99.0 %
Merck Index 15, 8789
IUPAC Name disodium hydrogen phosphate
Formula Weight 141.96
Percent Purity ≥99.0%
Identification Pass Test
Loss on Drying 0.2% max.
Packaging Poly Bottle
Decomposition Information Decomposes information: > 240°C
Chemical Name or Material Sodium Phosphate Dibasic Anhydrous
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.