missing translation for 'onlineSavingsMsg'
Learn More

Sodium phosphate dibasic dihydrate, 98+%, for analysis

Product Code. 12604957 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 kg
Packaging:
Plastic drum
Unit Size:
5kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
12604957 5 kg Plastic drum 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12604957 Supplier Thermo Scientific Acros Supplier No. 124990050
  • ONLINE EXCLUSIVE PRICE
    173.00 €
    167.00 € / 5kg
    Save 6.00 €
    3% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 10028-24-7 ∣ H5Na2O6P ∣ 177.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Melting Point 243.0°C to 245.0°C
CAS 10028-24-7
pH 8.4 to 9.6 (1% aq. soln.)
Physical Form Powder, Crystals and/or Chunks
Quantity 5 kg
Assay Percent Range +98%
Molecular Formula H5Na2O6P
Linear Formula Na2HPO4·2H2O
MDL Number MFCD00149182
Merck Index 15,8789
Synonym disodium hydrogen phosphate dihydrate, disodium phosphate dihydrate, sodium phosphate dibasic dihydrate, di-sodium hydrogen phosphate dihydrate, phosphoric acid, disodium salt, dihydrate, sodium hydrogenphosphate dihydrate, sodium phosphate, dibasic, dihydrate, disodium hydrogenphosphate dihydrate, dinatrii phosphas dihydricus, 2na.hpo4.2h2o
Solubility Information (5% in water) Clear colorless
InChI Key KDQPSPMLNJTZAL-UHFFFAOYSA-L
SMILES O.O.[Na+].[Na+].OP([O-])([O-])=O
IUPAC Name disodium;hydrogen phosphate;dihydrate
Molecular Weight (g/mol) 177.99
PubChem CID 6451167
Formula Weight 177.99
Percent Purity ≥98%
Grade Analytical
Loss on Drying 19.5% to 21.0% (1 g, 130°C)
Packaging Plastic drum
Chemical Name or Material Sodium phosphate dibasic dihydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.