missing translation for 'onlineSavingsMsg'
Learn More

Sodium phosphate dibasic dihydrate, 99+%, for HPLC

Product Code. 15204714 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic Bottle
Unit Size:
100g
2.5kg
25g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15204714 2.5 kg Plastic Bottle 2.5kg
10418862 25 g Glass bottle 25g
10005783 100 g Glass bottle 100g
10519273 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15204714 Supplier Thermo Scientific Acros Supplier No. 343810025
  • ONLINE EXCLUSIVE PRICE
    550.00 €
    512.00 € / 2.5kg
    Save 38.00 €
    6% Off
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 10028-24-7 | H5Na2O6P | 177.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10028-24-7
Molecular Formula H5Na2O6P
Molecular Weight (g/mol) 177.99
MDL Number MFCD00149182
InChI Key KDQPSPMLNJTZAL-UHFFFAOYSA-L
Synonym disodium hydrogen phosphate dihydrate, disodium phosphate dihydrate, sodium phosphate dibasic dihydrate, di-sodium hydrogen phosphate dihydrate, phosphoric acid, disodium salt, dihydrate, sodium hydrogenphosphate dihydrate, sodium phosphate, dibasic, dihydrate, disodium hydrogenphosphate dihydrate, dinatrii phosphas dihydricus, 2na.hpo4.2h2o
PubChem CID 6451167
IUPAC Name disodium;hydrogen phosphate;dihydrate
SMILES O.O.[Na+].[Na+].OP([O-])([O-])=O

Specifications

Melting Point 243.0°C to 245.0°C
Color White
pH 7.5 to 9.2 (5% soln. at 25°C)
Physical Form Crystalline Powder and/or Crystals
Quantity 2.5 kg
Packaging Plastic Bottle
Assay Percent Range 99+%
Linear Formula Na2HPO4·2H2O
Merck Index 15,8789
Solubility Information Solubility in water: 93g/L(20°C)
Formula Weight 177.99
Percent Purity ≥99%
Grade HPLC
Chemical Name or Material Sodium phosphate dibasic dihydrate, For HPLC
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.