missing translation for 'onlineSavingsMsg'
Learn More

Sodium phosphate dibasic dihydrate, Solstice

Product Code. 15696800
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15696800 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15696800 Supplier Honeywell-Fluka Supplier No. 71633250G
 Restricted Item
Request Bulk or Custom Format
Currently Unavailable: We have no information when this product will be available.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

for HPLC,≥98.5%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

CAS 10028-24-7
Physical Form Solid
Quantity 250 g
Molecular Formula H5Na2O6P
Linear Formula Na2HPO4 · 2H2O
MDL Number MFCD00149182
UN Number UN3077
Synonym disodium hydrogen phosphate dihydrate, disodium phosphate dihydrate, sodium phosphate dibasic dihydrate, di-sodium hydrogen phosphate dihydrate, phosphoric acid, disodium salt, dihydrate, sodium hydrogenphosphate dihydrate, sodium phosphate, dibasic, dihydrate, disodium hydrogenphosphate dihydrate, dinatrii phosphas dihydricus, 2na.hpo4.2h2o
InChI Key KDQPSPMLNJTZAL-UHFFFAOYSA-L
SMILES O.O.[Na+].[Na+].OP([O-])([O-])=O
IUPAC Name disodium;hydrogen phosphate;dihydrate
Molecular Weight (g/mol) 177.99
PubChem CID 6451167
Formula Weight 177.99g/mol
Percent Purity ≥98.5% (T)
Grade HPLC
Chemical Name or Material Sodium phosphate dibasic dihydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.