missing translation for 'onlineSavingsMsg'
Learn More

Sodium Phosphate Dibasic Dihydrate, puriss. p.a., Solstice

Product Code. 15623040
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
6 x 1 kg
Unit Size:
1kg
500g
5kg
Pack of 6
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15623040 5 kg 5kg
15613040 500 g 500g
15603040 1 kg 1kg
15633040 6 x 1 kg Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15623040 Supplier Honeywell-Fluka Supplier No. 304355KG
  • ONLINE EXCLUSIVE PRICE
    357.00 €
    322.08 € / 5kg
    Save 34.92 €
    9% Off
 Restricted Item
Estimated Shipment: 02-03-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Puriss. p.a., Reagent European Pharmacopoeia, 98.5-101.0% (calc. to the dried substance)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10028-24-7
Molecular Formula H5Na2O6P
Molecular Weight (g/mol) 177.99
MDL Number MFCD00149182
InChI Key KDQPSPMLNJTZAL-UHFFFAOYSA-L
Synonym disodium hydrogen phosphate dihydrate, disodium phosphate dihydrate, sodium phosphate dibasic dihydrate, di-sodium hydrogen phosphate dihydrate, phosphoric acid, disodium salt, dihydrate, sodium hydrogenphosphate dihydrate, sodium phosphate, dibasic, dihydrate, disodium hydrogenphosphate dihydrate, dinatrii phosphas dihydricus, 2na.hpo4.2h2o
PubChem CID 6451167
IUPAC Name disodium;hydrogen phosphate;dihydrate
SMILES O.O.[Na+].[Na+].OP([O-])([O-])=O

Specifications

Quantity 5 kg
Linear Formula Na2HPO4 · 2H2O
Formula Weight 177.99g/mol
Percent Purity 98.5 to 101% (calc. to the dried substance)
Grade puriss. p.a.
Chemical Name or Material Sodium Phosphate Dibasic Dihydrate, Reag. Ph. Eur.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.