missing translation for 'onlineSavingsMsg'
Learn More

Sodium phosphate dibasic dihydrate, specified accord. to the requir. of USP, Ph.Eur.

Product Code. 15760899 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
5 kg
Unit Size:
1kg
5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15760899 1 kg 1kg
15770899 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15760899 Supplier Thermo Scientific Acros Supplier No. 460790010
  • ONLINE EXCLUSIVE PRICE
    57.30 €
    53.70 € / 1kg
    Save 3.60 €
    6% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 10028-24-7 ∣ H5Na2O6P ∣ 177.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10028-24-7
Molecular Formula H5Na2O6P
Molecular Weight (g/mol) 177.99
MDL Number MFCD00149182
InChI Key KDQPSPMLNJTZAL-UHFFFAOYSA-L
IUPAC Name disodium dihydrate hydrogen phosphate
SMILES O.O.[Na+].[Na+].OP([O-])([O-])=O

Specifications

Melting Point 243.0°C to 245.0°C
Physical Form Powder, Crystals and/or Chunks
Quantity 1 kg
Packaging Glass Bottle
Linear Formula Na2HPO4·2 H2O
Solubility Information Solubility aq. soln.: 93 g/L(20°C).
Formula Weight 177.99
Grade Ungraded
Chemical Name or Material Sodium phosphate dibasic dihydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.