missing translation for 'onlineSavingsMsg'
Learn More

Sodium phosphate, dibasic dodecahydrate, 98.5%, for analysis

Product Code. 10227740 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
10 kg
Packaging:
Plastic bottle
Plastic drum
Unit Size:
10kg
1kg
2.5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10227740 1 kg Plastic bottle 1kg
10023350 2.5 kg Plastic bottle 2.5kg
10165263 10 kg Plastic drum 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10227740 Supplier Thermo Scientific Acros Supplier No. 271060010
  • ONLINE EXCLUSIVE PRICE
    50.80 €
    47.60 € / 1kg
    Save 3.20 €
    6% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 10039-32-4 | HNa2O4P·12H2O | 358.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10039-32-4
Molecular Formula HNa2O4P·12H2O
Molecular Weight (g/mol) 358.13
MDL Number MFCD00149181
InChI Key DGLRDKLJZLEJCY-UHFFFAOYSA-L
Synonym disodium phosphate dodecahydrate, disodium hydrogen phosphate dodecahydrate, phosphoric acid, disodium salt, dodecahydrate, di-sodium hydrogen phosphate dodecahydrate, sodium hydrogenphosphate dodecahydrate, unii-e1w4n241fo, sodium phosphate dibasic dodecahydrate, sodium phosphate, dibasic, dodecahydrate, disodium orthophosphate, dodecahydrate, sodium monohydrogen phosphate dodecahydrate 2:1:1:12
PubChem CID 61456
IUPAC Name disodium;hydrogen phosphate;dodecahydrate
SMILES O.O.O.O.O.O.O.O.O.O.O.O.OP(=O)([O-])[O-].[Na+].[Na+]

Specifications

Melting Point 35.0°C
Color White
pH 9 to 9.4 (5% aq. soln.)
Physical Form Crystals
Quantity 1 kg
Packaging Plastic bottle
Assay Percent Range 98.5%
Linear Formula Na2HPO4·12H2O
Merck Index 15, 8789
Solubility Information Solubility in water: 218g/L (20°C). Other solubilities: practically insoluble in alcohol
Formula Weight 358.13
Percent Purity 98.5%
Grade Analytical
Chemical Name or Material Sodium phosphate, dibasic dodecahydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.