missing translation for 'onlineSavingsMsg'
Learn More

Sodium phosphate, dibasic heptahydrate, 98+%, ACS reagent

Product Code. 10036020 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Plastic bottle
Unit Size:
1kg
500g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10036020 500 g Plastic bottle 500g
10297073 1 kg Plastic bottle 1kg
10175503 5 kg Plastic bottle 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10036020 Supplier Thermo Scientific Acros Supplier No. 424385000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7782-85-6 | H15Na2O11P | 268.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7782-85-6
Molecular Formula H15Na2O11P
Molecular Weight (g/mol) 268.06
MDL Number MFCD00149180
InChI Key LVXHNCUCBXIIPE-UHFFFAOYSA-L
Synonym sodium phosphate dibasic heptahydrate, unii-70wt22sf4b, sodium phosphate, dibasic, heptahydrate, sodium hydrogen phosphate heptahydrate, disodium hydrogen phosphate heptahydrate, axea paragraph signthornaae, 2na.hpo4.7h2o, sodium hydrogenphosphate heptahydrate, sodium phosphate dibasic, acs, disodium hydrogenphosphate heptahydrate
PubChem CID 6096963
IUPAC Name disodium phosphoric acid heptahydrate
SMILES O.[Na+].[Na+].OP([O-])([O-])=O

Specifications

Melting Point 48.0°C
Color White
pH 8.7 to 9.3 (5% soln. at 25°C)
Physical Form Crystals
Quantity 500 g
Assay Percent Range 98+%
Linear Formula Na2HPO4·7H2O
Merck Index 15, 8789
Formula Weight 268.07
Percent Purity ≥98%
Grade ACS Reagent
Packaging Plastic bottle
Decomposition Information >48°C
Density 1.67
Chemical Name or Material Sodium phosphate, dibasic heptahydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.