missing translation for 'onlineSavingsMsg'
Learn More

Sodium phosphate dibasic heptahydrate, Solstice

Product Code. 15618140 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
2.5 kg
6 x 500 g
4 x 2.5 kg
Unit Size:
100g
1kg
2.5kg
500g
Pack of 4
Pack of 6
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15618140 500 g 500g
15678130 100 g 100g
15688130 1 kg 1kg
15698130 2.5 kg 2.5kg
15628140 6 x 500 g Pack of 6
15608140 4 x 2.5 kg Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 15618140 Supplier Honeywell-Fluka Supplier No. S9390500G
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ACS reagent, 98.0-102.0%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7782-85-6
Molecular Formula H15Na2O11P
Molecular Weight (g/mol) 268.06
MDL Number MFCD00149180
InChI Key LVXHNCUCBXIIPE-UHFFFAOYSA-L
Synonym sodium phosphate dibasic heptahydrate, unii-70wt22sf4b, sodium phosphate, dibasic, heptahydrate, sodium hydrogen phosphate heptahydrate, disodium hydrogen phosphate heptahydrate, axea paragraph signthornaae, 2na.hpo4.7h2o, sodium hydrogenphosphate heptahydrate, sodium phosphate dibasic, acs, disodium hydrogenphosphate heptahydrate
PubChem CID 6096963
IUPAC Name disodium phosphoric acid heptahydrate
SMILES O.[Na+].[Na+].OP([O-])([O-])=O

Specifications

Quantity 500 g
Linear Formula Na2HPO4 · 7H2O
Formula Weight 268.07g/mol
Percent Purity 98 to 102%
Density 1.68g/mL at 25°C; Vapor Density: 4.9 (vs air)
Chemical Name or Material Sodium phosphate dibasic heptahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.