missing translation for 'onlineSavingsMsg'
Learn More

Sodium Phosphate, Dibasic, Heptahydrate, MP Biomedicals™

Product Code. 11494652 Shop All MP Biomedicals Products
Click to view available options
Quantity:
500 g
Unit Size:
500g
This item is not returnable. View return policy

Product Code. 11494652

Brand: MP Biomedicals 02191441.5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Widely used in molecular biology, biochemistry and chromatography. Sodium Phosphate, Dibasic, Heptahydrate, MP Biomedicals is a reagent with very high buffering capacity. It is highly hygroscopic, water soluble and useful (with Sodium Phosphate Monobasic)

Sodium phosphate dibasic heptahydrate is use in many applications such as:

  • Sequestrant, emulsifier and buffer in foods
  • As mordant in dyeing, for weighting silk and in tanning
  • In manufacturing of enamels, ceramics, detergents
  • As a fireproofing agent in soldering and brazing instead of borax
  • As a reagent and buffer in analytical chemistry, cathartic, laxative
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7782-85-6
Molecular Formula H15Na2O11P
Molecular Weight (g/mol) 268.06
MDL Number MFCD00149180
InChI Key LVXHNCUCBXIIPE-UHFFFAOYSA-L
Synonym sodium phosphate dibasic heptahydrate, unii-70wt22sf4b, sodium phosphate, dibasic, heptahydrate, sodium hydrogen phosphate heptahydrate, disodium hydrogen phosphate heptahydrate, axea paragraph signthornaae, 2na.hpo4.7h2o, sodium hydrogenphosphate heptahydrate, sodium phosphate dibasic, acs, disodium hydrogenphosphate heptahydrate
PubChem CID 6096963
IUPAC Name disodium phosphoric acid heptahydrate
SMILES O.[Na+].[Na+].OP([O-])([O-])=O

Specifications

Physical Form Crystalline Powder
Quantity 500 g
Solubility Information Soluble in water, but more soluble in hot water; insoluble in alcohol, ethanol.
Formula Weight 268.07
Percent Purity ≥98%
Grade ACS
compliance-icons
Product Identifier
  • Sodium Phosphate Dibasic, Heptahydrate, ACS
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Sodium Phosphate Dibasic, Heptahydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.