missing translation for 'onlineSavingsMsg'
Learn More

Sodium Phosphate Dibasic Heptahydrate, TRC

Product Code. 30422491
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30422491 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30422491 Supplier Toronto Research Chemicals Supplier No. TRCS667035500G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium Phosphate Dibasic Heptahydrate
CAS 7782-85-6
Molecular Formula HNa2O4P • 7(H2O)
Quantity 500 g
Synonym Disodium Hydrogen Phosphate Heptahydrate, Disodium Orthophosphate Heptahydrate, Disodium Phosphate Heptahydrate, Sodium Monohydrogen Phosphate Heptahydrate, Sodium Phosphate (Na2HPO4) Heptahydrate
InChI Formula InChI=1S/2Na.H3O4P.7H2O/c;;1-5(2,3)4;;;;;;;/h;;(H3,1,2,3,4);7*1H2/q2*+1;;;;;;;;/p-2
SMILES O.O.O.O.O.O.O.O=P([O-])([O-])O.[Na+].[Na+]
IUPAC Name disodium;hydrogen phosphate;heptahydrate
Molecular Weight (g/mol) 141.9671802
Formula Weight 268.0147
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.