missing translation for 'onlineSavingsMsg'
Learn More

Sodium phosphate dibasic, Solstice
BIOPHARMA

Product Code. 15615860
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15615860 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15615860 Supplier Honeywell-Fluka Supplier No. 568141KG
  • ONLINE EXCLUSIVE PRICE
    106.00 €
    95.67 € / 1kg
    Save 10.33 €
    9% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Tested according to Ph.Eur.

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

CAS 7558-79-4
Physical Form Powder
Quantity 1 kg
Molecular Formula HNa2O4P
Linear Formula Na2HPO4
MDL Number MFCD00003496
UN Number UN3077
Synonym disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt
InChI Key BNIILDVGGAEEIG-UHFFFAOYSA-L
SMILES [Na+].[Na+].OP([O-])([O-])=O
IUPAC Name disodium;hydrogen phosphate
Molecular Weight (g/mol) 141.96
PubChem CID 24203
ChEBI CHEBI:34683
Formula Weight 141.96g/mol
Percent Purity 100%
Grade Reagent
Chemical Name or Material Sodium phosphate dibasic
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.