missing translation for 'onlineSavingsMsg'
Learn More

Sodium Phosphate Dibasic, MP Biomedicals™

Product Code. 11495276 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
Unit Size:
100g
1kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11495276 1 kg 1kg
11410036 100 g 100g
11440246 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11495276 Supplier MP Biomedicals Supplier No. 0219980201
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Highly hygroscopic and water soluble. Sodium Phosphate Dibasic, MP Biomedicals is a sodium salt of phosphoric acid. This white powder is an important component of running buffer for denaturing gel electrophoresis.

Sodium phosphate dibasic anhydrous

  • Used for preparing phosphate buffer, which in turn is utilized in X-gal staining
  • Induces glomerular calcification as well as other nephrotic syndrome in Sprague-Dawley rats

Chemical Identifiers

CAS 7558-79-4
Molecular Formula HNa2O4P
Molecular Weight (g/mol) 141.96
MDL Number MFCD00003496
InChI Key BNIILDVGGAEEIG-UHFFFAOYSA-L
Synonym disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt
PubChem CID 24203
ChEBI CHEBI:34683
SMILES [Na+].[Na+].OP([O-])([O-])=O

Specifications

Physical Form Crystalline Powder
Quantity 1 kg
Solubility Information Freely soluble in water; insoluble in alcohol
IUPAC Name disodium hydrogen phosphate
Formula Weight 141.98
Percent Purity 98.0 to 100.5%
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.