missing translation for 'onlineSavingsMsg'
Learn More

Sodium Phosphate Dibasic, MP Biomedicals

Product Code. 11436621 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
250 g
500 g
1 kg
Grade:
98 to 102%
Molecular Biology
Unit Size:
1kg
250g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Grade unitSize
11436621 1 kg Molecular Biology 1kg
11416621 250 g Molecular Biology 250g
11426621 500 g Molecular Biology 500g
11455452 1 kg 98 to 102% 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11436621 Supplier MP Biomedicals Supplier No. 0219484991

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 7558-79-4
Molecular Formula HNa2O4P
Molecular Weight (g/mol) 141.96
MDL Number MFCD00003496
InChI Key BNIILDVGGAEEIG-UHFFFAOYSA-L
Synonym disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt
PubChem CID 24203
ChEBI CHEBI:34683
IUPAC Name disodium hydrogen phosphate
SMILES [Na+].[Na+].OP([O-])([O-])=O

Specifications

Quantity 1 kg
Formula Weight 141.96
Grade Molecular Biology
Chemical Name or Material Sodium Phosphate Dibasic, Anhydrous
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.