missing translation for 'onlineSavingsMsg'
Learn More

Sodium phosphate dibasic, Puriss., meets analytical spec. of BP, Ph. Eur., USP, FCC, E 339, anhydrous, 98-100.5% (calc. to the dried substance), Solstice
BIOPHARMA

Product Code. 15670750
Change view
Click to view available options
Quantity:
1 kg
5 kg
25 kg
4 x 5 kg
6 x 1 kg
Unit Size:
1kg
25kg
5kg
Pack of 4
Pack of 6
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15670750 1 kg 1kg
15600760 5 kg 5kg
15610760 6 x 1 kg Pack of 6
15690750 4 x 5 kg Pack of 4
15680750 25 kg 25kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 15670750 Supplier Honeywell-Fluka Supplier No. 042761KG
  • ONLINE EXCLUSIVE PRICE
    59.00 €
    53.00 € / 1kg
    Save 6.00 €
    10% Off
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Puriss., meets analytical specification of BP, Ph. Eur., USP, FCC, E 339, anhydrous, 98-100.5% (calc. to the dried substance)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7558-79-4
Molecular Formula HNa2O4P
Molecular Weight (g/mol) 141.96
MDL Number MFCD00003496
InChI Key BNIILDVGGAEEIG-UHFFFAOYSA-L
Synonym disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt
PubChem CID 24203
ChEBI CHEBI:34683
IUPAC Name disodium;hydrogen phosphate
SMILES [Na+].[Na+].OP([O-])([O-])=O

Specifications

Physical Form Anhydrous
Quantity 1 kg
Linear Formula Na2HPO4
Solubility Information Very Soluble in hot water, 8 parts soluble in water , insoluble in alcohol
Formula Weight 141.96g/mol
Percent Purity 98 to 100.5% (calc. to the dried substance)
Grade puriss.
Chemical Name or Material Sodium Phosphate Dibasic, Meets analytical specification of Ph.Eur.BP, USP, FCC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.