missing translation for 'onlineSavingsMsg'
Learn More

Sodium Phosphate Dibasic, (White, crystalline powder/ACS Reagent Grade), MP Biomedicals

Product Code. 11464652 Shop All MP Biomedicals Products
Change view
Click to view available options
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
11464652 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11464652 Supplier MP Biomedicals Supplier No. 0219144001
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 7558-79-4
Color White
Physical Form Crystalline Powder
Quantity 1 kg
Molecular Formula HNa2O4P
MDL Number MFCD00003496
Synonym disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt
InChI Key BNIILDVGGAEEIG-UHFFFAOYSA-L
SMILES [Na+].[Na+].OP([O-])([O-])=O
IUPAC Name disodium hydrogen phosphate
Molecular Weight (g/mol) 141.96
PubChem CID 24203
ChEBI CHEBI:34683
Formula Weight 141.96
Grade ACS Reagent
Chemical Name or Material Sodium Phosphate Dibasic
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.