missing translation for 'onlineSavingsMsg'
Learn More

Sodium phosphate dodecahydrate, 97%

Product Code. 11477687
Change view
Click to view available options
Quantity:
10,000 g
2500 g
500 g
Unit Size:
10000g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11477687 10,000 g 10000g
11457687 500 g 500g
11467687 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 11477687 Lieferant Thermo Scientific Alfa Aesar Lieferanten-Nr. A15713.0C
 Restricted Item
Estimated Shipment: 02-12-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

CAS: 10101-89-0 ∣ H24Na3O16P ∣ 380.119 g/mol

Sodium phosphate dodecahydrate is used in photographic developing, clarifying sugar, removing boiler scale, softening water and manufacturing of paper. They are added to many foods as an emulsifier to prevent oil separation. It can be utilized as a leavening agent. Some instances of these foods include the batter coating on breaded fish or chicken, and commercially baked cakes. Adding this item to food increases the shelf life of the food, maintaining the texture and visual aspect of the food.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications

Sodium phosphate dodecahydrate is used in photographic developing, clarifying sugar, removing boiler scale, softening water and manufacturing of paper. They are added to many foods as an emulsifier to prevent oil separation. It can be utilized as a leavening agent. Some instances of these foods include the batter coating on breaded fish or chicken, and commercially baked cakes. Adding this item to food increases the shelf life of the food, maintaining the texture and visual aspect of the food.

Solubility

Soluble in water. Insoluble in alcohol.

Notes

When stored under ordinary conditions, it may lose some water for crystallization. Incompatible with strong acids.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 10101-89-0
Molecular Formula H24Na3O16P
Molecular Weight (g/mol) 380.119
MDL Number MFCD00149198
InChI Key ASTWEMOBIXQPPV-UHFFFAOYSA-K
Synonym trisodium phosphate dodecahydrate, sodium phosphate dodecahydrate, sodium phosphate tribasic dodecahydrate, phosphoric acid, trisodium salt, dodecahydrate, unii-b70850qphr, ccris 7322, sodium phosphate, tribasic, dodecahydrate, trisodium dodecahydrate phosphate, phosphoric acid, trisodium salt, dodeahydrate, trisodium phosphate tert dodecahydrate
PubChem CID 61473
IUPAC Name trisodium;phosphate;dodecahydrate
SMILES O.O.O.O.O.O.O.O.O.O.O.O.[O-]P(=O)([O-])[O-].[Na+].[Na+].[Na+]

Spezifikation

Melting Point ∼75°C (decomposition)
pH 12
Physical Form Crystalline Powder, Needles or Crystals
Quantity 10,000 g
Odor Odorless
Density 1.62
Linear Formula Na3PO4·12H2O
UN Number UN3262
Merck Index 14,8662
Solubility Information Soluble in water. Insoluble in alcohol.
Formula Weight 380.12 (163.94 Anhydrous)
Percent Purity 97%
Grade Ungraded
Chemical Name or Material Sodium phosphate dodecahydrate
Mehr anzeigen Weniger anzeigen

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.