missing translation for 'onlineSavingsMsg'
Learn More

Sodium phosphate, monobasic, 98%, extra pure, anhydrous

Product Code. 12615157 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Plastic bottle
Unit Size:
1kg
5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
12615157 1 kg Plastic bottle 1kg
12625157 5 kg Plastic bottle 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12615157 Supplier Thermo Scientific Acros Supplier No. 448170010
  • ONLINE EXCLUSIVE PRICE
    54.50 €
    52.90 € / 1kg
    Save 1.60 €
    2% Off
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7558-80-7 ∣ H2NaO4P ∣ 119.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7558-80-7
Molecular Formula H2NaO4P
Molecular Weight (g/mol) 119.98
MDL Number MFCD00003527,MFCD00146206
InChI Key AJPJDKMHJJGVTQ-UHFFFAOYSA-M
Synonym monosodium phosphate, sodium dihydrogen phosphate, sodium dihydrogenorthophosphate, sodium phosphate monobasic, sodium acid phosphate, sodium phosphate, monobasic, monosodium dihydrogen orthophosphate, sodium phosphate monobasic anhydrous, monosodium orthophosphate, monobasic sodium phosphate
PubChem CID 23672064
ChEBI CHEBI:37585
IUPAC Name sodium;dihydrogen phosphate
SMILES [Na+].OP(O)([O-])=O

Specifications

Physical Form Crystalline Powder
Quantity 1 kg
Assay Percent Range 98%
Linear Formula NaH2PO4
Merck Index 15,879
Solubility Information Solubility in water: 850g/L (20°C).
Formula Weight 119.98
Percent Purity 98%
Grade Extra Pure
Packaging Plastic bottle
Chemical Name or Material Sodium phosphate, monobasic
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.