missing translation for 'onlineSavingsMsg'
Learn More

Melford Sodium Phosphate, Monobasic, Anhydrous

Product Code. p-200086982
Click to view available options
Quantity:
500 g
1 kg
5 kg
Unit Size:
1kg
500g
5kg
This item is not returnable. View return policy

Product Code. 30018793

Brand: Melford S231855000.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sodium Phosphate, Monobasic, Anhydrous is a chemical compound that plays a significant role in the life science research.

It is also known as monosodium phosphate or sodium dihydrogen phosphate. The anhydrous designation indicates that it is the form of the compound without any water molecules.

In the life science field, it is primarily utilized as a buffering agent and as a source of phosphate ions. Buffers are substances that help maintain a stable pH in biological and biochemical processes, ensuring optimal conditions for enzymatic reactions and other cellular activities.

Sodium Phosphate, Monobasic, Anhydrous is often used in laboratory settings for the preparation of various biological reagents, cell culture media, and buffer solutions. It can be combined with other phosphate compounds to create buffer systems with specific pH ranges and buffering capacities, making it suitable for a wide range of experimental conditions

Moreover, the compound is used in molecular biology techniques like DNA and protein electrophoresis, as it provides the necessary pH control for separation and analysis of biomolecules. It is also employed in the preparation of phosphate-based saline solutions.

Specifications

Chemical Name or Material Sodium Phosphate, Monobasic, Anhydrous
CAS 7558-80-7
Quantity 5 kg
Molecular Formula NaH2PO4
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.