missing translation for 'onlineSavingsMsg'
Learn More

Sodium Phosphate, Monobasic, Dihydrate, BAKER ANALYZED™ Reagent, J.T. Baker™

Product Code. 15578174 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15578174 500 g Poly Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15578174 Supplier Avantor J.T.Baker Supplier No. 381901
  • ONLINE EXCLUSIVE PRICE
    335.00 €
    265.30 € / 500g
    Save 69.70 €
    20% Off
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High quality chemicals for laboratory and specialized industrial use

Specifications

CAS 13472-35-0
Quantity 500 g
Molecular Formula H6NaO6P
Synonym sodium dihydrogen phosphate dihydrate, sodium phosphate monobasic dihydrate, monosodium phosphate dihydrate, sodium phosphate, monobasic, dihydrate, phosphoric acid, monosodium salt, dihydrate, sodium dihydrogenphosphate dihydrate, sodium phosphate nah2po4 dihydrate, monosodium dihydrogen phosphate dihydrate, sodium dihydrate dihydrogen phosphate, acmc-20ajoh
InChI Key VBJGJHBYWREJQD-UHFFFAOYSA-M
SMILES O.O.OP(=O)(O)[O-].[Na+]
IUPAC Name sodium;dihydrogen phosphate;dihydrate
Molecular Weight (g/mol) 156.006
PubChem CID 23673460
Formula Weight 155.99
Grade BAKER ANALYZED™ Reagent
Packaging Poly Bottle
Specific Gravity 1L = 1.92kg
Chemical Name or Material Sodium Phosphate, Monobasic, Dihydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.