missing translation for 'onlineSavingsMsg'
Learn More

Sodium phosphate, monobasic dihydrate, for HPLC

Product Code. 16697012 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
Packaging:
glass bottle
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
16697012 250 g glass bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16697012 Supplier Thermo Scientific Acros Supplier No. 446222500
  • ONLINE EXCLUSIVE PRICE
    57.20 €
    55.50 € / 250g
    Save 1.70 €
    2% Off
View all promotions
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 13472-35-0 | H6NaO6P | 156.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Melting Point 60°C
CAS 13472-35-0
Color White
pH 4.0 to 4.5
Physical Form Crystals
Quantity 250 g
Molecular Formula H6NaO6P
Linear Formula NaH2PO4.2H2O
MDL Number MFCD00149209
Synonym Sodium dihydrogen phosphate dihydrate
Solubility Information soluble aq. soln., practically insoluble in alcohol
InChI Key VBJGJHBYWREJQD-UHFFFAOYSA-M
SMILES O.O.[Na+].OP(O)([O-])=O
IUPAC Name sodium dihydrate dihydrogen phosphate
Molecular Weight (g/mol) 156.01
Formula Weight 156.01
Percent Purity 98.5 to 101.5 % (Titration acid)
Grade HPLC
Packaging glass bottle
Appearance White Crystals
Chemical Name or Material Sodium phosphate, monobasic dihydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.