missing translation for 'onlineSavingsMsg'
Learn More

Sodium phosphate, monobasic monohydrate, 98+%, ACS reagent

Product Code. 10566992 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
500 g
2.5 kg
10 kg
Packaging:
Plastic bottle
Plastic drum
Unit Size:
10kg
2.5kg
25g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10566992 25 g Plastic bottle 25g
10657961 500 g Plastic bottle 500g
10708922 2.5 kg Plastic bottle 2.5kg
11924291 10 kg Plastic drum 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10566992 Supplier Thermo Scientific Acros Supplier No. 424390250
  • ONLINE EXCLUSIVE PRICE
    28.35 €
    27.50 € / 25g
    Save 0.85 €
    2% Off
View all promotions
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 10049-21-5 | H5NaO5P | 139.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10049-21-5
Molecular Formula H5NaO5P
Molecular Weight (g/mol) 139.00
MDL Number MFCD00149208
InChI Key BBMHARZCALWXSL-UHFFFAOYSA-N
Synonym sodium dihydrogen phosphate monohydrate, sodium phosphate monobasic monohydrate, monosodium phosphate monohydrate, unii-593yog76rn, phosphoric acid, monosodium salt, monohydrate, sodium phosphate monobasic hydrate, sodium dihydrogen phosphate hydrate, sodium dihydrogenphosphate monohydrate, sodium hydrate dihydrogen phosphate, pubchem12707
PubChem CID 516949
IUPAC Name sodium phosphoric acid hydrate
SMILES O.[Na+].OP(O)(O)=O

Specifications

Melting Point 100.0°C
Color White
pH 4.1 to 4.5 (5% soln. at 25°C)
Physical Form Crystalline Powder and/or Crystals
Quantity 25 g
Assay Percent Range 98+%
Linear Formula NaH2PO4·H2O
Fieser 01,395
Merck Index 15,879
Solubility Information Solubility in water: 1103g/L (20°C). Other solubilities: practically insoluble in alcohol
Formula Weight 137.99
Percent Purity ≥98%
Grade ACS Reagent
Packaging Plastic bottle
Chemical Name or Material Sodium phosphate, monobasic monohydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.