missing translation for 'onlineSavingsMsg'
Learn More

Sodium phosphate monobasic monohydrate, ACS Reagent, Solstice

Product Code. 15618150
Change view
Click to view available options
Quantity:
25 g
250 g
500 g
1 kg
3 kg
6 x 1 kg
Unit Size:
1kg
250g
25g
3kg
500g
Pack of 6
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15618150 1 kg 1kg
15638150 25 g 25g
15628150 250 g 250g
15658150 500 g 500g
15648150 3 kg 3kg
15668150 6 x 1 kg Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 15618150 Supplier Honeywell-Fluka Supplier No. S96381KG
  • ONLINE EXCLUSIVE PRICE
    121.00 €
    109.05 € / 1kg
    Save 11.95 €
    9% Off
 Restricted Item
Estimated Shipment: 26-03-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

ACS Reagent,≥98%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 10049-21-5
Molecular Formula H5NaO5P
Molecular Weight (g/mol) 139.00
MDL Number MFCD00149208
InChI Key BBMHARZCALWXSL-UHFFFAOYSA-N
Synonym sodium dihydrogen phosphate monohydrate, sodium phosphate monobasic monohydrate, monosodium phosphate monohydrate, unii-593yog76rn, phosphoric acid, monosodium salt, monohydrate, sodium phosphate monobasic hydrate, sodium dihydrogen phosphate hydrate, sodium dihydrogenphosphate monohydrate, sodium hydrate dihydrogen phosphate, pubchem12707
PubChem CID 516949
SMILES O.[Na+].OP(O)(O)=O

Tekniske data

Quantity 1 kg
Linear Formula NaH2PO4 · H2O
UN Number NONH for all modes of transport
IUPAC Name sodium;dihydrogen phosphate;hydrate
Formula Weight 137.99g/mol
Percent Purity 98 to 102%, ≥98%
Grade ACS Reagent
Chemical Name or Material Sodium phosphate monobasic monohydrate
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.