missing translation for 'onlineSavingsMsg'
Learn More

Sodium phosphate monobasic monohydrate, ACS Reagent, Solstice

Product Code. 15668150 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
25 g
250 g
500 g
1 kg
3 kg
6 x 1 kg
Unit Size:
1kg
250g
25g
3kg
500g
Pack of 6
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15668150 6 x 1 kg Pack of 6
15638150 25 g 25g
15628150 250 g 250g
15658150 500 g 500g
15618150 1 kg 1kg
15648150 3 kg 3kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 15668150 Supplier Honeywell-Fluka Supplier No. S96386X1KG
 Restricted Item
Request Bulk or Custom Format
Currently Unavailable: We have no information when this product will be available.
View alternative products
This item is not returnable. View return policy

ACS Reagent,≥98%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10049-21-5
Molecular Formula H5NaO5P
Molecular Weight (g/mol) 139.00
MDL Number MFCD00149208
InChI Key BBMHARZCALWXSL-UHFFFAOYSA-N
Synonym sodium dihydrogen phosphate monohydrate, sodium phosphate monobasic monohydrate, monosodium phosphate monohydrate, unii-593yog76rn, phosphoric acid, monosodium salt, monohydrate, sodium phosphate monobasic hydrate, sodium dihydrogen phosphate hydrate, sodium dihydrogenphosphate monohydrate, sodium hydrate dihydrogen phosphate, pubchem12707
PubChem CID 516949
SMILES O.[Na+].OP(O)(O)=O

Specifications

Quantity 6 x 1 kg
Linear Formula NaH2PO4 · H2O
UN Number NONH for all modes of transport
IUPAC Name sodium;dihydrogen phosphate;hydrate
Formula Weight 137.99g/mol
Percent Purity 98 to 102%, ≥98%
Grade ACS Reagent
Chemical Name or Material Sodium phosphate monobasic monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.