missing translation for 'onlineSavingsMsg'
Learn More

Sodium Phosphate, Monobasic, Monohydrate, Crystal, U.S.P. - F.C.C., J.T. Baker™

Product Code. 15588174 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Wide Mouth Amber Glass Bottle
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15588174 500 g Wide Mouth Amber Glass Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15588174 Supplier Avantor J.T.Baker Supplier No. 382001
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity crystalline powder meeting USP and FCC standards for laboratory use.

Specifications

CAS 10049-21-5
Physical Form Crystals
Quantity 500 g
Molecular Formula H5NaO5P
MDL Number MFCD00149208
Synonym sodium dihydrogen phosphate monohydrate, sodium phosphate monobasic monohydrate, monosodium phosphate monohydrate, unii-593yog76rn, phosphoric acid, monosodium salt, monohydrate, sodium phosphate monobasic hydrate, sodium dihydrogen phosphate hydrate, sodium dihydrogenphosphate monohydrate, sodium hydrate dihydrogen phosphate, pubchem12707
InChI Key BBMHARZCALWXSL-UHFFFAOYSA-N
SMILES O.[Na+].OP(O)(O)=O
IUPAC Name sodium;dihydrogen phosphate;hydrate
Molecular Weight (g/mol) 139.00
PubChem CID 516949
Formula Weight 137.99
Percent Purity ≥98%
Grade FCC/USP
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 2.04kg
Chemical Name or Material Sodium Phosphate, Monobasic, Monohydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.