missing translation for 'onlineSavingsMsg'
Learn More

Sodium Phosphate, Monobasic, Monohydrate, Crystal, Ultrapure Bioreagent, J.T. Baker™

Product Code. 15539670 Shop All J.T.Baker Products
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
Unit Size:
500g
Each
This item is not returnable. View return policy

Product Code. 15539670

Brand: Avantor J.T.Baker 401101

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

    For Liquid Chromatography and Molecular Biology applications

Chemical Identifiers

CAS 10049-21-5
Molecular Formula H5NaO5P
Molecular Weight (g/mol) 139.00
MDL Number MFCD00149208
InChI Key BBMHARZCALWXSL-UHFFFAOYSA-N
Synonym sodium dihydrogen phosphate monohydrate, sodium phosphate monobasic monohydrate, monosodium phosphate monohydrate, unii-593yog76rn, phosphoric acid, monosodium salt, monohydrate, sodium phosphate monobasic hydrate, sodium dihydrogen phosphate hydrate, sodium dihydrogenphosphate monohydrate, sodium hydrate dihydrogen phosphate, pubchem12707
PubChem CID 516949
IUPAC Name sodium;dihydrogen phosphate;hydrate
SMILES O.[Na+].OP(O)(O)=O

Specifications

Physical Form Crystals
Quantity 500 g
Formula Weight 137.99
Grade Ultrapure Bioreagent
Packaging Poly Bottle
Specific Gravity 1L = 2.04kg
Chemical Name or Material Sodium Phosphate, Monobasic, Monohydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.