missing translation for 'onlineSavingsMsg'
Learn More

Sodium Phosphate, Monobasic, Monohydrate, Crystal, Ultrapure Bioreagent, J.T. Baker™

Product Code. 15558204 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
Unit Size:
500g
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15558204 2.5 kg Poly Bottle Each
15539670 500 g Poly Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15558204 Supplier Avantor J.T.Baker Supplier No. 401105
 Restricted Item
Estimated Shipment: 16-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

    For Liquid Chromatography and Molecular Biology applications

Chemical Identifiers

CAS 10049-21-5
Molecular Formula H5NaO5P
Molecular Weight (g/mol) 139.00
MDL Number MFCD00149208
InChI Key BBMHARZCALWXSL-UHFFFAOYSA-N
Synonym sodium dihydrogen phosphate monohydrate, sodium phosphate monobasic monohydrate, monosodium phosphate monohydrate, unii-593yog76rn, phosphoric acid, monosodium salt, monohydrate, sodium phosphate monobasic hydrate, sodium dihydrogen phosphate hydrate, sodium dihydrogenphosphate monohydrate, sodium hydrate dihydrogen phosphate, pubchem12707
PubChem CID 516949
IUPAC Name sodium;dihydrogen phosphate;hydrate
SMILES O.[Na+].OP(O)(O)=O

Specifications

Physical Form Crystals
Quantity 2.5 kg
Formula Weight 137.99
Grade Ultrapure Bioreagent
Packaging Poly Bottle
Specific Gravity 1L = 2.04kg
Chemical Name or Material Sodium Phosphate, Monobasic, Monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.