missing translation for 'onlineSavingsMsg'
Learn More

Sodium phosphate monobasic monohydrate, For ACS analysis, 98.0 to 102.0%, MP Biomedicals™

Product Code. 11424662 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11424662 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11424662 Supplier MP Biomedicals Supplier No. 02191442.1
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 10049-21-5
Color White
Physical Form Crystalline Powder
Quantity 100 g
Assay Percent Range 98.0 to 102.0%
Molecular Formula H5NaO5P
MDL Number MFCD00149208
Synonym sodium dihydrogen phosphate monohydrate, sodium phosphate monobasic monohydrate, monosodium phosphate monohydrate, unii-593yog76rn, phosphoric acid, monosodium salt, monohydrate, sodium phosphate monobasic hydrate, sodium dihydrogen phosphate hydrate, sodium dihydrogenphosphate monohydrate, sodium hydrate dihydrogen phosphate, pubchem12707
InChI Key BBMHARZCALWXSL-UHFFFAOYSA-N
SMILES O.[Na+].OP(O)(O)=O
IUPAC Name sodium phosphoric acid hydrate
Molecular Weight (g/mol) 139.00
PubChem CID 516949
Formula Weight 137.99
Chemical Name or Material Sodium phosphate monobasic monohydrate, For ACS analysis
Show More Show Less
compliance-icons
Product Identifier
  • Sodium Phosphate Monobasic Monohydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Sodium Phosphate Monobasic Monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.